15(S)-HETE ELISA Kit
Catalog # ABCAAB133035-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Competitive
- Format:Pre-coated
- Target protein:15(S)-HETE
- Description:15(S)-HETE ELISA kit
- Sample type:Urine, plasma, cell culture supernatant, serum
- Detection method:Colorimetric
- Detection range:78,1 - 20000 pg/ml
- Storage temperature:4 °C
- Sensitivity:69,21 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
15(S)-HETE ELISA kit is a Competitive ELISA kit for the measurement of 15(S)-HETE in Urine, plasma, cell culture supernatant, Serum samples.