Adrenaline ELISA Kit
Catalog # ABCAAB287788-96
Supplier: Abcam
CAS Number:
Specifications
- Pk:96 Tests
- Assay Type:Competitive
- Target protein:Adrenaline (Epinephrine)
- Description:Adrenaline ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Adrenaline ELISA Kit ab287788 is used to quantify Adrenaline with a sensitivity of 2,1 pg/ml. Competitive ELISA Kit.