Human GCNT1 ELISA Kit
Catalog # ABCAAB313963-96TST
Supplier: Abcam
CAS Number:
Specifications
- Description:Human GCNT1 ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Human GCNT1 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human GCNT1 in Human in serum, plasma, cell culture supernatant samples.