Natalizumab ELISA Kit
ABCAAB282908-1X96
: Abcam
:
- Description:Natalizumab ELISA kit
- Tests per kit:96 tests
Natalizumab ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Natalizumab in Human in plasma, serum samples.
Natalizumab ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Natalizumab in Human in plasma, serum samples.