Human Acetylcholinesterase ELISA Kit - Extracellular
Supplier: Abcam
New Product
Certificates
About this item
Human Acetylcholinesterase ELISA kit - Extracellular is a single-wash 90-min Simplestep used to quantify Human Acetylcholinesterase with a sensitivity of 20,314 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput. Validated on a number of sample types including cerebrospinal fluid (CSF).
Specifications
- Storage temperature:+4 ℃