Mouse IL-19 Antibody Pair - BSA and Azide free
ABCAAB256723-10X96
: Abcam
:
- Description:Mouse IL-19 Antibody Pair - BSA and Azide free
- Tests per kit:10×96 tests
Mouse IL-19 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Mouse IL-19.