Order Entry
Export
ContactUsLinkComponent
Anti-Cyclin E1 Rabbit Monoclonal Antibody [clone: EPR194] (Alexa Fluor® 488)
Anti-Cyclin E1 Rabbit Monoclonal Antibody [clone: EPR194] (Alexa Fluor® 488)
Catalog # ABCAAB319576-100UL
Supplier:  Abcam
New Product
Anti-Cyclin E1 Rabbit Monoclonal Antibody [clone: EPR194] (Alexa Fluor® 488)
Catalog # ABCAAB319576-100UL
Supplier:  Abcam
New Product

Specifications

  • Seq:
    1286
  • Size:
    100 µl
  • Pack type:
    Vial

Specifications

About this item

Alexa Fluor® 488 Rabbit monoclonal [EPR194] to Cyclin E1.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Cyclin E1
Clonality: Monoclonal
Clone: EPR194
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: