Anti-DAP12 Rabbit Monoclonal Antibody [clone: EPR24244-76] (Alexa Fluor® 647)
Supplier: Abcam
New Product
Certificates
About this item
Alexa Fluor® 647 Rabbit monoclonal [EPR24244-76] to DAP12.
Type: Primary
Antigen: DAP12
Clonality: Monoclonal
Clone: EPR24244-76
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse