Anti-Cytokeratin 17 Rabbit Monoclonal Antibody [clone: EPR1624Y] (Alexa Fluor® 594)
Catalog # ABCAAB310484-100UL
Supplier: Abcam
Specifications
- Seq:1286
- Size:100 µl
- Pack type:Vial
Specifications
About this item
Alexa Fluor® 594 Rabbit monoclonal [EPR1624Y] to Cytokeratin 17.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Cytokeratin 17
Clonality: Monoclonal
Clone: EPR1624Y
Conjugation: Alexa Fluor® 594
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: