Anti-SCA2 Rabbit Monoclonal Antibody [clone: EPR23630-49]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR23630-49] to SCA2.
Type: Primary
Antigen: SCA2
Clonality: Monoclonal
Clone: EPR23630-49
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat