Anti-NCR1 Rabbit Monoclonal Antibody [clone: EPR23097-35]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR23097-35] to NCR1.
Type: Primary
Antigen: NCR1
Clonality: Monoclonal
Clone: EPR23097-35
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat