Anti-IL-33 Rabbit Monoclonal Antibody [clone: EPR17831]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR17831] to IL-33.
Type: Primary
Antigen: IL-33
Clonality: Monoclonal
Clone: EPR17831
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat