Anti-CD36 Rabbit Monoclonal Antibody [clone: EPR22509-40]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR22509-40] to CD36.
Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: EPR22509-40
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat; Mouse