Anti-Histone H2B (acetyl K20) Rabbit Monoclonal Antibody [clone: EPR859]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR859] to Histone H2B (acetyl K20) - ChIP Grade.
Type: Primary
Antigen: Histone H2B
Clonality: Monoclonal
Clone: EPR859
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat