Anti-Cytokeratin 17 Mouse Monoclonal Antibody [clone: KRT17/778]
ABCAAB212553-100
: Abcam
- Size:100 µg
- Pack type:Vial
Mouse monoclonal [KRT17/778] to Cytokeratin 17 - BSA and Azide free.
Type: Primary
Antigen: Cytokeratin 17
Clonality: Monoclonal
Clone: KRT17/778
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human