Anti-Mannose Receptor Rabbit Monoclonal Antibody [clone: EPR22489-7]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR22489-7] to Mannose Receptor - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Mannose Receptor
Clonality: Monoclonal
Clone: EPR22489-7
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCAAB254471-10
- ABCAAB254471-1
- Antigen symbol:
- Mannose Receptor
- Mannose Receptor
- Antigen name:
- CD206
- CD206
- Antigen synonyms:
- CLEC13D|CLEC13DL|MRC1L1|MRC1|Macrophage mannose receptor 1|MMR|C-type lectin domain family 13 member D|C-type lectin domain family 13 member D-like|Human mannose receptor|Macrophage mannose receptor 1-like protein 1|hMR
- CLEC13D|CLEC13DL|MRC1L1|MRC1|Macrophage mannose receptor 1|MMR|C-type lectin domain family 13 member D|C-type lectin domain family 13 member D-like|Human mannose receptor|Macrophage mannose receptor 1-like protein 1|hMR
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR22489-7
- EPR22489-7
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular weight:
- 166012 Da
- 166012 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage temperature:
- Store at 4 °C. Do not freeze.
- Store at 4 °C. Do not freeze.
- Shipping temperature:
- 4 °C
- 4 °C
- Tested applications:
- IHC-P
- IHC-P