Anti-CCR7 Rabbit Monoclonal Antibody [clone: EPR23192-57]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR23192-57] to CCR7 - BSA and Azide free.
Type: Primary
Antigen: CCR7
Clonality: Monoclonal
Clone: EPR23192-57
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat