Anti-Folate Binding Protein/FBP Rabbit Monoclonal Antibody [clone: EPR23387-276] (PE)
Catalog # ABCAAB275200-100
Supplier: Abcam
Specifications
- Size:100 µl
- Pack type:Vial
Specifications
About this item
PE Rabbit monoclonal [EPR23387-276] to Folate Binding Protein/FBP.
Type: Primary
Antigen: Folate Binding Protein/FBP
Clonality: Monoclonal
Clone: EPR23387-276
Conjugation: PE
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human