Anti-MMP9 Rabbit Monoclonal Antibody [clone: EPR22154-211]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR22154-211] to MMP9 - BSA and Azide free (Detector).
Type: Primary
Antigen: MMP9
Clonality: Monoclonal
Clone: EPR22154-211
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Monkey