Anti-Syndecan-1 Rabbit Monoclonal Antibody [clone: EPR22809-30]
: Abcam
Rabbit monoclonal [EPR22809-30] to Syndecan-1 - BSA and Azide free (Capture).
Type: Primary
Antigen: Syndecan-1
Clonality: Monoclonal
Clone: EPR22809-30
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human