Anti-pan Cytokeratin Mouse Monoclonal Antibody [clone: C-11] (Alexa Fluor® 555)
Catalog # ABCAAB279324-100
Supplier: Abcam
Specifications
- Size:100 µl
- Pack type:Vial
Specifications
About this item
Alexa Fluor® 555 Mouse monoclonal [C-11] to pan Cytokeratin.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: pan Cytokeratin
Clonality: Monoclonal
Clone: C-11
Conjugation: Alexa Fluor® 555
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human