Anti-DYRK1A Rabbit Monoclonal Antibody [clone: EPR24132-61]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR24132-61] to DYRK1A.
Type: Primary
Antigen: DYRK1A
Clonality: Monoclonal
Clone: EPR24132-61
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat