Anti-MAP2 Rabbit Monoclonal Antibody [clone: EPR22641-16]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR22641-16] to MAP2.
Type: Primary
Antigen: MAP2
Clonality: Monoclonal
Clone: EPR22641-16
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat