Anti-TIM 1 Rabbit Monoclonal Antibody [clone: EPR22650-115]
: Abcam
Rabbit monoclonal [EPR22650-115] to TIM 1 - BSA and Azide free (Capture).
Type: Primary
Antigen: TIM 1
Clonality: Monoclonal
Clone: EPR22650-115
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human