Order Entry
Export
ContactUsLinkComponent
Anti-StAR Rabbit Monoclonal Antibody [clone: HL1362]
Anti-StAR Rabbit Monoclonal Antibody [clone: HL1362]
Catalog # ABCAAB307801-100
Supplier:  Abcam
undefined
Anti-StAR Rabbit Monoclonal Antibody [clone: HL1362]
Catalog # ABCAAB307801-100
Supplier:  Abcam

Specifications

  • Seq:
    1286
  • Size:
    100 µl
  • Pack type:
    Vial

Specifications

About this item

Rabbit monoclonal [HL1362] to StAR - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: StAR
Clonality: Monoclonal
Clone: HL1362
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse