Anti-beta Amyloid 1-42 Rabbit Monoclonal Antibody [clone: mOC64]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [mOC64] to beta Amyloid 1-42.
Type: Primary
Antigen: beta Amyloid 1-42
Clonality: Monoclonal
Clone: mOC64
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat