Anti-Sez6 Rabbit Monoclonal Antibody [clone: EPR28518-55]
: Abcam
New Product
Rabbit monoclonal [EPR28518-55] to Sez6.
Type: Primary
Antigen: Sez6
Clonality: Monoclonal
Clone: EPR28518-55
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat