Anti-CD163 Rabbit Monoclonal Antibody [clone: EPR14643-36] (Alexa Fluor® 647)
Catalog # ABCAAB218294-100
Supplier: Abcam
Specifications
- Size:100 µl
- Pack type:Vial
Specifications
About this item
Alexa Fluor® 647 Rabbit monoclonal [EPR14643-36] to CD163 - C-terminal.
Type: Primary
Antigen: CD163
Clonality: Monoclonal
Clone: EPR14643-36
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human