Anti-Tau Alzheimer’s Disease Mouse Monoclonal Antibody [clone: GT-38]
Catalog # ABCAAB246808-100
Supplier: Abcam
Specifications
- Size:100 µg
- Pack type:Vial
Specifications
About this item
Mouse monoclonal [GT-38] to Tau Alzheimer’s Disease - Conformation-Specific.
Type: Primary
Antigen: Tau Alzheimer’s Disease
Clonality: Monoclonal
Clone: GT-38
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human