Anti-Cyclin E1 Rabbit Monoclonal Antibody [clone: EP435E] (Alexa Fluor® 647)
ABCAAB194069-100
: Abcam
- Size:100 µl
- Pack type:Vial
Alexa Fluor® 647 Rabbit monoclonal [EP435E] to Cyclin E1.
Type: Primary
Antigen: Cyclin E1
Clonality: Monoclonal
Clone: EP435E
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human