Anti-IL-20 Rabbit Monoclonal Antibody [clone: EPR20756-1]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR20756-1] to IL-20 - BSA and Azide free (Capture).
Type: Primary
Antigen: IL-20
Clonality: Monoclonal
Clone: EPR20756-1
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse