Anti-alpha 1 Antitrypsin Rabbit Monoclonal Antibody [clone: EPR17087-50]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR17087-50] to alpha 1 Antitrypsin.
Type: Primary
Antigen: alpha 1 Antitrypsin
Clonality: Monoclonal
Clone: EPR17087-50
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human