Anti-Connexin 43 / GJA1 Rabbit Monoclonal Antibody [clone: EPR21153]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR21153] to Connexin 43 / GJA1.
Type: Primary
Antigen: Connexin 43 / GJA1
Clonality: Monoclonal
Clone: EPR21153
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human