Anti-RELN Rabbit Monoclonal Antibody [clone: EPR26278-30]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR26278-30] to Reelin.
Type: Primary
Antigen: RELN
Clonality: Monoclonal
Clone: EPR26278-30
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat