Anti-MEK1 (phospho T292) Rabbit Monoclonal Antibody [clone: EPR2365Y]
: Abcam
Rabbit monoclonal [EPR2365Y] to MEK1 (phospho T292) - BSA and Azide free.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: MEK1
Clonality: Monoclonal
Clone: EPR2365Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ABCAAB247465-1
- ABCAAB247465-100
- Antigen symbol:
- MEK1
- MEK1
- Antigen name:
- PRKMK1
- PRKMK1
- Antigen synonyms:
- MAP2K1|Dual specificity mitogen-activated protein kinase kinase 1|MAP kinase kinase 1|MAPKK 1|MKK1|ERK activator kinase 1|MAPK/ERK kinase 1|MEK 1
- MAP2K1|Dual specificity mitogen-activated protein kinase kinase 1|MAP kinase kinase 1|MAPKK 1|MKK1|ERK activator kinase 1|MAPK/ERK kinase 1|MEK 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR2365Y
- EPR2365Y
- Form:
- Liquid
- Liquid