Anti-TOMM20 Rabbit Monoclonal Antibody [clone: EPR15581-54] (Alexa Fluor® 555)
ABCAAB221292-100
: Abcam
- Size:100 µl
- Pack type:Vial
Alexa Fluor® 555 Rabbit monoclonal [EPR15581-54] to TOMM20 - Mitochondrial Marker.
Type: Primary
Antigen: TOMM20
Clonality: Monoclonal
Clone: EPR15581-54
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human