Anti-IL-1 beta Rabbit Monoclonal Antibody [clone: EPR18815-27]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR18815-27] to IL-1 beta - BSA and Azide free (Capture).
Type: Primary
Antigen: IL-1 beta
Clonality: Monoclonal
Clone: EPR18815-27
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Monkey