Anti-Cathepsin D Mouse Monoclonal Antibody [clone: CTD-19]
Catalog # ABCAAB6313-50
Supplier: Abcam
Specifications
- Size:50 µl
- Pack type:Vial
Specifications
About this item
Mouse monoclonal [CTD-19] to Cathepsin D.
Type: Primary
Antigen: Cathepsin D
Clonality: Monoclonal
Clone: CTD-19
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human