Anti-CDKN2A/p16INK4a Rabbit Monoclonal Antibody [clone: EPR24167-43]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR24167-43] to CDKN2A/p16INK4a.
Type: Primary
Antigen: CDKN2A/p16INK4a
Clonality: Monoclonal
Clone: EPR24167-43
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human