Anti-VE Cadherin Mouse Monoclonal Antibody [clone: 55-7H1] (Alexa Fluor® 488)
Catalog # ABCAAB272345-100
Supplier: Abcam
Specifications
- Size:100 Tests
- Pack type:Vial
Specifications
About this item
Alexa Fluor® 488 Mouse monoclonal [55-7H1] to VE Cadherin.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: VE Cadherin
Clonality: Monoclonal
Clone: 55-7H1
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human