Anti-CD36 Rabbit Monoclonal Antibody [clone: EPR22512-58]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR22512-58] to CD36 - BSA and Azide free.
Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: EPR22512-58
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat