Anti-Lamin A + Lamin C Rabbit Monoclonal Antibody [clone: EPR4100] (Alexa Fluor® 594)
Catalog # ABCAAB215324-100
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:LMN1
- Antigen symbol:Lamin A + Lamin C
- Clonality:Monoclonal
- Clone:EPR4100
- Conjugation:Alexa Fluor® 594
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Form:Liquid
- Antigen synonyms:LMNA|Prelamin-A/C
- Molecular weight:74139 Da
- Storage temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze / thaw cycle. Store in the dark.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information.
- Tested applications:ICC/IF
- Purification:Protein A Affinity purification
- Size:100 µl
- Pack type:Vial
Specifications
About this item
Alexa Fluor® 594 Rabbit monoclonal [EPR4100] to Lamin A + Lamin C - Nuclear Envelope Marker.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Lamin A + Lamin C
Clonality: Monoclonal
Clone: EPR4100
Conjugation: Alexa Fluor® 594
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human