Anti-Kappa light chain Mouse Monoclonal Antibody [clone: OX12] (Alexa Fluor® 488)
ABCAAB256420-100
: Abcam
- Size:100 µl
- Pack type:Vial
Alexa Fluor® 488 Mouse monoclonal [OX12] to Kappa light chain.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Kappa light chain
Clonality: Monoclonal
Clone: OX12
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat