Anti-Src (phospho Y419) Rabbit Monoclonal Antibody [clone: EPR17734]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR17734] to Src (phospho Y419) - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Src
Clonality: Monoclonal
Clone: EPR17734
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Specifications
- Catalog No:
- ABCAAB209615-1
- ABCAAB209615-100
- Antigen symbol:
- Src
- Src
- Antigen name:
- SRC1
- SRC1
- Antigen synonyms:
- Proto-oncogene tyrosine-protein kinase Src|Proto-oncogene c-Src|pp60c-src|p60-Src
- Proto-oncogene tyrosine-protein kinase Src|Proto-oncogene c-Src|pp60c-src|p60-Src
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR17734
- EPR17734
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Modification:
- phospho Y419
- phospho Y419
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular weight:
- 59835 Da
- 59835 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage temperature:
- Store at 4 °C. Do not freeze.
- Store at 4 °C. Do not freeze.
- Shipping temperature:
- 4 °C
- 4 °C
- Tested applications:
- Dot, Flow Cyt (Intra)
- Dot, Flow Cyt (Intra)