Anti-Fast Myosin Skeletal Heavy chain Mouse Monoclonal Antibody [clone: MY-32]
ABCAAB51263-50
: Abcam
- Size:50 µl
- Pack type:Vial
Mouse monoclonal [MY-32] to Fast Myosin Skeletal Heavy chain.
Type: Primary
Antigen: Fast Myosin Skeletal Heavy chain
Clonality: Monoclonal
Clone: MY-32
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rabbit