Order Entry
Export
ContactUsLinkComponent
Anti-Coiled-coil domain-containing protein 113 Mouse Monoclonal Antibody [clone: 18C8BC7AD10]
Anti-Coiled-coil domain-containing protein 113 Mouse Monoclonal Antibody [clone: 18C8BC7AD10]
Catalog # ABCAAB221849-100
Supplier:  Abcam
undefined
Anti-Coiled-coil domain-containing protein 113 Mouse Monoclonal Antibody [clone: 18C8BC7AD10]
Catalog # ABCAAB221849-100
Supplier:  Abcam

Specifications

  • Seq:
    1286
  • Size:
    100 µg
  • Pack type:
    Vial

Specifications

About this item

Mouse IgG2c, kappa monoclonal [18C8BC7AD10] - Isotype Control - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Control
Antigen: Coiled-coil domain-containing protein 113
Clonality: Monoclonal
Clone: 18C8BC7AD10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2c
Reactivity: