Anti-KMT1E / SETDB1 Rabbit Monoclonal Antibody [clone: EPR24218-43]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR24218-43] to KMT1E / SETDB1.
Type: Primary
Antigen: KMT1E / SETDB1
Clonality: Monoclonal
Clone: EPR24218-43
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human