Anti-SPAM1 Rabbit Monoclonal Antibody [clone: EPR22906-57]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR22906-57] to SPAM1 - BSA and Azide free.
Type: Primary
Antigen: SPAM1
Clonality: Monoclonal
Clone: EPR22906-57
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human