Anti-CD4 Rabbit Monoclonal Antibody [clone: EPR20339-289]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR20339-289] to CD4 - BSA and Azide free (Detector).
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: EPR20339-289
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human