Order Entry
Export
ContactUsLinkComponent
 
Anti-BMP4 Rabbit Monoclonal Antibody [clone: EPR18235-128]
 :  Abcam
 
undefined
Anti-BMP4 Rabbit Monoclonal Antibody [clone: EPR18235-128]
 :  Abcam

 

  • Catalog No:
  • ABCAAB244725-1
  • ABCAAB244725-100
  • Antigen symbol:
  • BMP4
  • BMP4
  • Antigen name:
  • BMP2B
  • BMP2B
  • Antigen synonyms:
  • DVR4|Bone morphogenetic protein 4|BMP-4|Bone morphogenetic protein 2B|BMP-2B
  • DVR4|Bone morphogenetic protein 4|BMP-4|Bone morphogenetic protein 2B|BMP-2B
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR18235-128
  • EPR18235-128
  • Form:
  • Liquid
  • Liquid